(苯磺酰)乙腈 ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
P0799-25G
P0799-5G
TCIAP0799-25GEA
0
CNY
TCIAP0799-25G
TCIAP0799-5G
(苯磺酰)乙腈 ≥98.0% (通过GC)
(苯磺酰)乙腈
分子式:
C₈H₇NO₂S |
MDL编号:
MFCD00007550 CAS号: 7605-28-9 EINECS: 231-515-0 UN号: 3439 ADR: 6.1,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...