Pregnenolone ≥98.0% (by GC)

供应商: TCI

P0786-5G P0786-25G
TCIAP0786-5GEA 0 CNY
TCIAP0786-5G TCIAP0786-25G
Pregnenolone ≥98.0% (by GC)
Pregnenolone
分子式: C₂₁H₃₂O₂
MDL编号: MFCD00003628
CAS号: 145-13-1
EINECS: 205-647-4
Merck Index: 14,07733

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 187.0 to 192.0 deg-C
Specific rotation [a]20/D +26.0 to +29.0 deg (C=1,EtOH)
Solubility in hot Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR