Propionylcholine-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
供应商: TCI
P0517-1G
P0517-25G
TCIAP0517-1GEA
0
CNY
TCIAP0517-1G
TCIAP0517-25G
Propionylcholine-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
Propionylcholine-p-toluenesulphonate
分子式:
C₁₅H₂₅NO₅S 分子量: 331.43 g/mol |
MDL编号:
MFCD00059975 CAS号: 1866-13-3 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...