孕烯诺龙乙酸酯 ≥99.0% (通过HPLC)

供应商: TCI

P0477-10G P0477-1G
TCIAP0477-10GEA 0 CNY
TCIAP0477-10G TCIAP0477-1G
孕烯诺龙乙酸酯 ≥99.0% (通过HPLC)
孕烯诺龙乙酸酯
分子式: C₂₃H₃₄O₃
MDL编号: MFCD00003627
CAS号: 1778-02-5
EINECS: 217-212-6
Merck Index: 14,07733

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 149.0 to 152.0 deg-C
Specific rotation [a]20/D +17.0 to +21.0 deg (C=1, EtOH)
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR