Polyvinylpyrrolidone, M.W. ∼ 40000 K 30

供应商: TCI

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P0472-25G P0472-500G
TCIAP0472-25GEA 0 CNY
TCIAP0472-25G TCIAP0472-500G
Polyvinylpyrrolidone, M.W. ∼ 40000 K 30
Polyvinylpyrrolidone
分子式: (C₆H₉NO)n
MDL编号: MFCD01076626
CAS号: 9003-39-8
Merck Index: 14,07697

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Total Nitrogen 12.0 to 12.8 w/w% (calcd.on anh.substance)
Water max. 7.0 %
K value 26.0 to 34.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR