Piperazine adipate ≥98.0% (by titrimetric analysis)
供应商: TCI
P0448-25G
P0448-500G
TCIAP0448-25GEA
0
CNY
TCIAP0448-25G
TCIAP0448-500G
Piperazine adipate ≥98.0% (by titrimetric analysis)
Piperazine adipate
分子式:
C₄H₁₀N₂ 分子量: 232.28 g/mol |
MDL编号:
MFCD00050650 CAS号: 142-88-1 EINECS: 205-569-0 Merck Index: 14,07465 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...