Disodium phthalate ≥95.0% (通过滴定分析)

供应商: TCI

P0400-25G P0400-500G
TCIAP0400-25GEA 0 CNY
TCIAP0400-25G TCIAP0400-500G
Disodium phthalate ≥95.0% (通过滴定分析)
Disodium phthalate
分子式: C₈H₆O₄
MDL编号: MFCD00050736
CAS号: 15968-01-1
EINECS: 240-106-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR