(R)-(−)-Phenylephrine hydrochloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: L(-)-Phenylephrine hydrochloride, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol hydrochloride, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol hydrochloride, (R)-Phenylephrine hydrochloride, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol hydrochloride, R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

P0398-25G P0398-5G
TCIAP0398-25GEA 0 CNY
TCIAP0398-25G TCIAP0398-5G
(R)-(−)-Phenylephrine hydrochloride ≥98.0% (by HPLC, titration analysis)
(R)-(−)-Phenylephrine hydrochloride
分子式: C₉H₁₃NO₂·HCl
MDL编号: MFCD00012605
CAS号: 61-76-7
EINECS: 200-517-3
Merck Index: 14,07286

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 142.0 to 145.0 deg-C
Specific rotation [a]20/D -44.0 to -47.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR