L-Proline methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: H-Pro-OMe.HCl, H-Pro-OMe·HCl

P0342-25G P0342-5G
TCIAP0342-25GEA 0 CNY
TCIAP0342-25G TCIAP0342-5G
L-Proline methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Proline methyl ester hydrochloride
分子式: C₆H₁₁NO₂·HCl
分子量: 165.6 g/mol
沸点: 55°C
存储条件: Refrigerator
MDL编号: MFCD00012708
CAS号: 2133-40-6
EINECS: 218-363-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 75.0 deg-C
Specific rotation [a]20/D -30.0 to -33.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR