Methyl cinnamate 98%

供应商: Thermo Fisher Scientific

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

279760050 279765000
ACRO279760050EA 0 CNY
ACRO279760050 ACRO279765000
Methyl cinnamate 98%
Methyl cinnamate
分子式: C₁₀H₁₀O₂
分子量: 162.19 g/mol
溶点: 33.5…36 °C
闪点: >110 °C
MDL编号: MFCD00008458
CAS号: 103-26-4
EINECS: 203-093-8

现在订购

产品技术规格

Appearance White to light yellow Fused crystalline mass
Infrared spectrum Conforms
Melting point 33.5 to 36 °C
GC ≥97.5 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR