Pectin (from citrus)

供应商: TCI

Synonyms: Poly-D-galacturonic acid methyl ester

P0024-500G P0024-25G
TCIAP0024-500GEA 0 CNY
TCIAP0024-500G TCIAP0024-25G
Pectin (from citrus)
Pectin
MDL编号: MFCD00081838
CAS号: 9000-69-5
EINECS: 232-553-0
Merck Index: 14,07063

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Drying loss max. 10.0 %
Galactronic Acid min. 58.0 %
Methoxyl Group min. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR