N-(4-Oxocyclohexyl)phthalimide ≥98.0% (by GC, total nitrogen)

供应商: TCI

Synonyms: 4-Phthalimidocyclohexanone, 2-(4-Oxocyclohexyl)isoindoline-1,3-dione

O0365-25G O0365-5G
TCIAO0365-25GEA 0 CNY
TCIAO0365-25G TCIAO0365-5G
N-(4-Oxocyclohexyl)phthalimide ≥98.0% (by GC, total nitrogen)
N-(4-Oxocyclohexyl)phthalimide
分子式: C₁₄H₁₃NO₃
分子量: 243.26 g/mol
MDL编号: MFCD00158659
CAS号: 104618-32-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR