Omeprazole ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

O0359-25G O0359-5G
TCIAO0359-25GEA 0 CNY
TCIAO0359-25G TCIAO0359-5G
Omeprazole ≥98.0% (by HPLC, titration analysis)
Omeprazole
分子式: C₁₇H₁₉N₃O₃S
分子量: 345.42 g/mol
存储条件: Refrigerator
MDL编号: MFCD00083192
CAS号: 73590-58-6
Merck Index: 14,06845

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR