Triethyl orthodichloroacetate

供应商: TCI
Warning

Synonyms: 1,1,1-Triethoxy-2,2-dichloroethane, Orthodichloroacetic Acid Triethyl Ester

O0274-1G O0274-5G
TCIAO0274-1GEA 0 CNY
TCIAO0274-1G TCIAO0274-5G
Triethyl orthodichloroacetate
Triethyl orthodichloroacetate
分子式: C₈H₁₆Cl₂O₃
沸点: 195°C
存储条件: Refrigerator
MDL编号: MFCD01861293
CAS号: 54567-92-9

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR