1-氧杂-2-氧代-3-硫杂中氮茚氯盐 ≥98.0% (by total nitrogen basis) for source of alkyl radical

供应商: TCI
Warning

Synonyms: 氯化 2-氧代[1,4,2]噁噻唑并[2,3-a]吡啶鎓

O0270-1G O0270-5G
TCIAO0270-1GEA 0 CNY
TCIAO0270-1G TCIAO0270-5G
1-氧杂-2-氧代-3-硫杂中氮茚氯盐 ≥98.0% (by total nitrogen basis) for source of alkyl radical
1-氧杂-2-氧代-3-硫杂中氮茚氯盐
分子式: C₆H₄ClNO₂S
分子量: 189.61 g/mol
MDL编号: MFCD00134271
CAS号: 89025-51-4

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR