trans-9-Octadecenol ≥99.0% (通过GC)

供应商: TCI

O0228-1G O0228-5G
TCIAO0228-1GEA 0 CNY
TCIAO0228-1G TCIAO0228-5G
trans-9-Octadecenol ≥99.0% (通过GC)
trans-9-Octadecenol
分子式: C₁₈H₃₆O
沸点: 333°C
存储条件: Refrigerator
MDL编号: MFCD00063218
CAS号: 506-42-3
EINECS: 208-037-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 35.0 to 39.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR