1,1′-草酰基二咪唑 ≥95.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 草酰基二咪唑

O0200-1G O0200-5G
TCIAO0200-1GEA 0 CNY
TCIAO0200-1G TCIAO0200-5G
1,1′-草酰基二咪唑 ≥95.0% (by titrimetric analysis)
1,1′-草酰基二咪唑
分子式: C₈H₆N₄O₂
分子量: 190.16 g/mol
存储条件: Refrigerator
MDL编号: MFCD00043067
CAS号: 18637-83-7

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR