Oleyl Phosphate (Mono- and Di- ester mixture)

供应商: TCI
Warning

Synonyms: Phosphoric acid Oleyl ester

O0146-25ML O0146-500ML
TCIAO0146-25MLEA 0 CNY
TCIAO0146-25ML TCIAO0146-500ML
Oleyl Phosphate (Mono- and Di- ester mixture)
Oleyl Phosphate (Mono- and Di- ester mixture)
MDL编号: MFCD00135103
CAS号: 37310-83-1
EINECS: 253-455-4

现在订购

产品技术规格

Appearance White or Colorless to Brown powder to lump to clear liquid
Assay of mono ester 37.0 to 50.0 %
Assay of Diester 50.0 to 63.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR