n-Octylsuccinic anhydride ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
O0049-25G
O0049-250G
TCIAO0049-25GEA
0
CNY
TCIAO0049-25G
TCIAO0049-250G
n-Octylsuccinic anhydride ≥98.0% (by GC, titration analysis)
n-Octylsuccinic anhydride
分子式:
C₁₂H₂₀O₃ |
MDL编号:
MFCD00014553 CAS号: 4200-92-4 EINECS: 224-102-1 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...