Isobutyl n-octanoate ≥99.0% (by GC)

供应商: TCI

Synonyms: n-Octanoic Acid Isobutyl Ester

O0032-25ML O0032-500ML
TCIAO0032-25MLEA 0 CNY
TCIAO0032-25ML TCIAO0032-500ML
Isobutyl n-octanoate ≥99.0% (by GC)
Isobutyl n-octanoate
分子式: C₁₂H₂₄O₂
闪点: 108°C
MDL编号: MFCD00059444
CAS号: 5461-06-3
EINECS: 226-744-8

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR