Nonanal dimethyl acetal ≥97.0% (by GC)
供应商: TCI
Synonyms:
Pelargonaldehyde Dimethyl Acetal, 1,1-Dimethoxynonane, Nonyl Aldehyde Dimethyl Acetal
N0915-5G
N0915-25G
TCIAN0915-5GEA
0
CNY
TCIAN0915-5G
TCIAN0915-25G
Nonanal dimethyl acetal ≥97.0% (by GC)
Nonanal dimethyl acetal
分子式:
C₁₁H₂₄O₂ 沸点: 88°C |
MDL编号:
MFCD00036646 CAS号: 18824-63-0 EINECS: 242-603-3 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...