Norfloxacin ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
N0817-25G
N0817-5G
TCIAN0817-25GEA
0
CNY
TCIAN0817-25G
TCIAN0817-5G
Norfloxacin ≥98.0% (by HPLC, titration analysis)
Norfloxacin
分子式:
C₁₆H₁₈FN₃O₃ 分子量: 319.34 g/mol 溶点: 221°C |
MDL编号:
MFCD00079532 CAS号: 70458-96-7 EINECS: 274-614-4 Merck Index: 14,06700 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...