Neu5Ac-ɑ(2-3)Gal-β-MP Glycoside ≥95.0% (by HPLC)

供应商: TCI

Synonyms: 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP

N0791-10MG N0791-50MG
TCIAN0791-10MGEA 0 CNY
TCIAN0791-10MG TCIAN0791-50MG
Neu5Ac-ɑ(2-3)Gal-β-MP Glycoside ≥95.0% (by HPLC)
Neu5Ac-alpha(2-3)Gal-beta-MP Glycoside
分子式: C₂₄H₃₅NO₁₅
分子量: 577.54 g/mol
存储条件: Freezer
CAS号: 159922-54-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR