2-萘乙醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
alpha-Methyl-2-naphthalenemethanol, 2-(1-Hydroxyethyl)naphthalene
N0788-25G
N0788-5G
TCIAN0788-25GEA
0
CNY
TCIAN0788-25G
TCIAN0788-5G
2-萘乙醇 ≥98.0% (by GC)
2-萘乙醇
分子式:
C₁₂H₁₂O 分子量: 172.23 g/mol 沸点: 170°C |
MDL编号:
MFCD00004111 CAS号: 7228-47-9 EINECS: 230-630-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...