2-(2-Nitrovinyl)thiophene ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-(2-Nitroethenyl)thiophene
N0787-25G
N0787-5G
TCIAN0787-25GEA
0
CNY
TCIAN0787-25G
TCIAN0787-5G
2-(2-Nitrovinyl)thiophene ≥98.0% (by GC)
2-(2-Nitrovinyl)thiophene
分子式:
C₆H₅NO₂S 分子量: 155.17 g/mol |
MDL编号:
MFCD00069434 CAS号: 874-84-0 EINECS: 212-867-4 |
产品技术规格
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...