3-Noradamantanamine hydrochloride ≥98.0% (通过总氮和滴定分析)
供应商: TCI
|
Warning
|
N0778-1G
N0778-5G
TCIAN0778-1GEA
0
CNY
TCIAN0778-1G
TCIAN0778-5G
3-Noradamantanamine hydrochloride ≥98.0% (通过总氮和滴定分析)
3-Noradamantanamine hydrochloride
分子式:
C₉H₁₅N 溶点: 350°C |
MDL编号:
MFCD00213499 CAS号: 86128-83-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...