3-(1-Naphthyl)-L-alanine hydrochloride ≥97.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
(S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
N0773-1G
N0773-5G
TCIAN0773-1GEA
0
CNY
TCIAN0773-1G
TCIAN0773-5G
3-(1-Naphthyl)-L-alanine hydrochloride ≥97.0% (by HPLC, titration analysis)
3-(1-Naphthyl)-L-alanine hydrochloride
分子式:
C₁₃H₁₃NO₂ 分子量: 251.71 g/mol |
CAS号:
122745-10-2 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 97.0 % |
Specific rotation [a]20/D | -6.0 to -10.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...