1,5-萘二磺酸 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
Armstrong 酸
N0763-25G
N0763-500G
TCIAN0763-25GEA
0
CNY
TCIAN0763-25G
TCIAN0763-500G
1,5-萘二磺酸 ≥98.0% (by HPLC, titration analysis)
1,5-萘二磺酸
分子式:
C₁₀H₁₆O₁₀S₂ |
MDL编号:
MFCD00149265 CAS号: 211366-30-2 UN号: 3261 ADR: 8,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 18.0 to 23.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...