8-Nonen-1-ol ≥98.0% (by GC)

供应商: TCI

N0650-25G N0650-5G
TCIAN0650-25GEA 0 CNY
TCIAN0650-25G TCIAN0650-5G
8-Nonen-1-ol ≥98.0% (by GC)
8-Nonen-1-ol
分子式: C₉H₁₈O
分子量: 142.24 g/mol
沸点: 134°C
MDL编号: MFCD00798062
CAS号: 13038-21-6

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR