5-降冰片烯-2,2-二甲醇 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: NDM

N0636-1G N0636-5G
TCIAN0636-1GEA 0 CNY
TCIAN0636-1G TCIAN0636-5G
5-降冰片烯-2,2-二甲醇 ≥98.0% (by GC)
5-降冰片烯-2,2-二甲醇
分子式: C₉H₁₄O₂
分子量: 154.21 g/mol
MDL编号: MFCD00167595
CAS号: 6707-12-6

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 111.0 to 115.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR