4-硝基苯甲酰乙酸乙酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: 3-(4-Nitrophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Nitrobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Nitrophenyl)-3-oxopropionate

N0513-25G N0513-5G
TCIAN0513-25GEA 0 CNY
TCIAN0513-25G TCIAN0513-5G
4-硝基苯甲酰乙酸乙酯 ≥98.0% (by GC)
4-硝基苯甲酰乙酸乙酯
分子式: C₁₁H₁₁NO₅
分子量: 237.21 g/mol
MDL编号: MFCD00007357
CAS号: 838-57-3
EINECS: 212-656-7

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR