4-Nitropyrrole-2-carboxylic acid hydrate ≥99.0% (by HPLC, titration analysis)

供应商: TCI

N0509-1G N0509-5G
TCIAN0509-1GEA 0 CNY
TCIAN0509-1G TCIAN0509-5G
4-Nitropyrrole-2-carboxylic acid hydrate ≥99.0% (by HPLC, titration analysis)
4-Nitropyrrole-2-carboxylic acid hydrate
分子式: C₅H₄N₂O₄
溶点: 217°C
MDL编号: MFCD00059928
CAS号: 5930-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9eae9eaabd8d753c839915c43e8895166655a5c&vid=beb9d92b8b5982a731837f47d44e6209cb6f373e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 % (calcd.on anh.substance)
Solubility in Methanol almost transparency
Water max. 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR