炔诺酮乙酸酯 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 17α-乙炔基-17β-羟基-19-去甲-4-雄甾烯-3-酮 17-乙酸酯, 去甲基脱氢羟孕酮 17-乙酸酯, 17α-乙炔基-19-去甲睾酮 17β-乙酸酯, 17-羟基-19-去甲-17α-孕甾-4-烯-20-炔基-3-酮 乙酸酯

N0450-1G N0450-100MG
TCIAN0450-1GEA 0 CNY
TCIAN0450-1G TCIAN0450-100MG
炔诺酮乙酸酯 ≥98.0% (by HPLC, titration analysis)
炔诺酮乙酸酯
分子式: C₂₂H₂₈O₃
分子量: 340.46 g/mol
MDL编号: MFCD04039850
CAS号: 51-98-9
EINECS: 200-132-0
Merck Index: 14,06697

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 165.0 deg-C
Specific rotation [a]20/D -33.0 to -36.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR