2-Isobutoxynaphthalene ≥98.0% (by GC)
供应商: TCI
Synonyms:
Isobutyl 2-Naphthyl Ether
N0433-25G
N0433-500G
TCIAN0433-25GEA
0
CNY
TCIAN0433-25G
TCIAN0433-500G
2-Isobutoxynaphthalene ≥98.0% (by GC)
2-Isobutoxynaphthalene
分子式:
C₁₄H₁₆O 分子量: 200.28 g/mol 闪点: 68°C |
MDL编号:
MFCD00046467 CAS号: 2173-57-1 EINECS: 218-529-2 |
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...