(±)-1-(1-萘基)乙醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
(±)-α-甲基-1-萘甲醇
N0395-1G
N0395-5G
TCIAN0395-1GEA
0
CNY
TCIAN0395-1G
TCIAN0395-5G
(±)-1-(1-萘基)乙醇 ≥98.0% (by GC)
(±)-1-(1-萘基)乙醇
分子式:
C₁₂H₁₂O 分子量: 172.23 g/mol 沸点: 133°C |
MDL编号:
MFCD00016806 CAS号: 1517-72-2 EINECS: 216-172-7 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 65.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...