1-Nitro-2-naphthol ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Hydroxy-1-nitronaphthalene, 1-Nitronaphthalen-2-ol
N0370-1G
N0370-5G
TCIAN0370-1GEA
0
CNY
TCIAN0370-1G
TCIAN0370-5G
1-Nitro-2-naphthol ≥98.0% (by GC, titration analysis)
1-Nitro-2-naphthol
分子式:
C₁₀H₇NO₃ |
MDL编号:
MFCD00021461 CAS号: 550-60-7 EINECS: 208-984-5 Merck Index: 14,06614 |
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...