叔壬基硫醇,异构体混合物 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 叔壬基硫醇

N0299-25ML N0299-500ML
TCIAN0299-25MLEA 0 CNY
TCIAN0299-25ML TCIAN0299-500ML
叔壬基硫醇,异构体混合物 ≥97.0% (by titrimetric analysis)
叔壬基硫醇,异构体混合物
分子式: C₉H₂₀S
分子量: 160.32 g/mol
闪点: 64°C
MDL编号: MFCD00059963
CAS号: 25360-10-5
EINECS: 246-896-9

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) many component mixture
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR