Sodium-3-nitrosalicylate ≥99.0% (通过滴定分析)

供应商: TCI
Warning

N0258-25G N0258-5G
TCIAN0258-25GEA 0 CNY
TCIAN0258-25G TCIAN0258-5G
Sodium-3-nitrosalicylate ≥99.0% (通过滴定分析)
Sodium-3-nitrosalicylate
分子式: C₇H₅NO₅
MDL编号: MFCD00070529
CAS号: 164915-53-1

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR