2-Nitrophenol sodium salt ≥99.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Sodium 2-Nitrophenol

N0222-25G N0222-500G
TCIAN0222-25GEA 0 CNY
TCIAN0222-25G TCIAN0222-500G
2-Nitrophenol sodium salt ≥99.0% (by HPLC, titration analysis)
2-Nitrophenol sodium salt
分子式: C₆H₅NO₃
分子量: 161.09 g/mol
MDL编号: MFCD00065178
CAS号: 824-39-5
EINECS: 212-527-5

现在订购

产品技术规格

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR