5-Nitrofluorescein (isomer I) ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 5-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, 3',6'-Dihydroxy-5-nitro-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

N0199-1G N0199-5G
TCIAN0199-1GEA 0 CNY
TCIAN0199-1G TCIAN0199-5G
5-Nitrofluorescein (isomer I) ≥98.0% (by titrimetric analysis)
5-Nitrofluorescein (isomer I)
分子式: C₂₀H₁₁NO₇
分子量: 377.31 g/mol
MDL编号: MFCD00135108
CAS号: 3326-35-0

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in NaOH (100g/L) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR