Mevastatin ≥98.0% (by HPLC)

供应商: TCI

Synonyms: (1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate, Compactin

M2275-1G M2275-5G
TCIAM2275-1GEA 0 CNY
TCIAM2275-1G TCIAM2275-5G
Mevastatin ≥98.0% (by HPLC)
Mevastatin
分子式: C₂₃H₃₄O₅
分子量: 390.52 g/mol
存储条件: Refrigerator
MDL编号: MFCD05662341
CAS号: 73573-88-3
Merck Index: 14,06164

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 151.0 to 155.0 deg-C
Specific rotation [a]20/D +280 to +295 deg (C=0.4, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR