Methyl-9-formylnonanoate ≥95.0% (by GC)
供应商: TCI
Synonyms:
9-Formylnonanoic Acid Methyl Ester
M2263-1G
M2263-5G
TCIAM2263-1GEA
0
CNY
TCIAM2263-1G
TCIAM2263-5G
Methyl-9-formylnonanoate ≥95.0% (by GC)
Methyl-9-formylnonanoate
分子式:
C₁₁H₂₀O₃ 分子量: 200.28 g/mol 沸点: 121°C 存储条件: Refrigerator |
CAS号:
14811-73-5 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...