Molindone hydrochloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Danger

M2252-100MG M2252-1G
TCIAM2252-100MGEA 0 CNY
TCIAM2252-100MG TCIAM2252-1G
Molindone hydrochloride ≥98.0% (通过HPLC,滴定分析)
Molindone hydrochloride
分子式: C₁₆H₂₄N₂O₂
MDL编号: MFCD01718304
CAS号: 15622-65-8
UN号: 2811
ADR: 6.1,III
Merck Index: 14,06233

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR