Marbofloxacin ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,3,4]oxadiazino[6,5,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,3,4]oxadiazino[6,5,4-ij]quinoline-6-carboxylic acid

M2240-1G M2240-5G
TCIAM2240-1GEA 0 CNY
TCIAM2240-1G TCIAM2240-5G
Marbofloxacin ≥98.0% (by HPLC, titration analysis)
Marbofloxacin
分子式: C₁₇H₁₉FN₄O₄
分子量: 362.36 g/mol
溶点: 269°C
MDL编号: MFCD00864820
CAS号: 115550-35-1
Merck Index: 14,05750

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR