7-Methoxycoumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 7-Methoxy-2-oxo-2H-chromene-3-carboxylic acid

M2233-1G M2233-100MG
TCIAM2233-1GEA 0 CNY
TCIAM2233-1G TCIAM2233-100MG
7-Methoxycoumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)
7-Methoxycoumarin-3-carboxylic acid
分子式: C₁₁H₈O₅
分子量: 220.18 g/mol
存储条件: Freezer
MDL编号: MFCD00452770
CAS号: 20300-59-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR