1-甲基-1H-吡唑-4-羧酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 1-甲基-4-吡唑甲酸

M2224-5G M2224-1G
TCIAM2224-5GEA 0 CNY
TCIAM2224-5G TCIAM2224-1G
1-甲基-1H-吡唑-4-羧酸 ≥98.0% (by GC, titration analysis)
1-甲基-1H-吡唑-4-羧酸
分子式: C₅H₆N₂O₂
分子量: 126.1100 g/mol
MDL编号: MFCD00159641
CAS号: 5952-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58331c438386982c4f4a5aa486b755149ff0d4b2&vid=58331c438386982c4f4a5aa486b755149ff0d4b2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 205.0 to 209.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR