甲基吡咯-2-羧酸酯 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
M2160-25G
M2160-5G
TCIAM2160-25GEA
0
CNY
TCIAM2160-25G
TCIAM2160-5G
甲基吡咯-2-羧酸酯 ≥98.0% (通过GC)
甲基吡咯-2-羧酸酯
分子式:
C₆H₇NO₂ |
MDL编号:
MFCD00817048 CAS号: 1193-62-0 |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...