3-Methoxybutyl thioglycolate ≥97.0% (by GC, titration analysis)

供应商: TCI

Synonyms: 3-Methoxybutyl 2-mercaptoacetate, 3-Methoxybutyl Mercaptoacetate, Mercaptoacetic Acid 3-Methoxybutyl Ester, Thioglycolic Acid 3-Methoxybutyl Ester

M2060-25G M2060-500G
TCIAM2060-25GEA 0 CNY
TCIAM2060-25G TCIAM2060-500G
3-Methoxybutyl thioglycolate ≥97.0% (by GC, titration analysis)
3-Methoxybutyl thioglycolate
分子式: C₇H₁₄O₃S
分子量: 178.25 g/mol
MDL编号: MFCD04038084
CAS号: 27431-39-6

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR