2-Methoxy-1-naphthalenemethanol ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-(Hydroxymethyl)-2-methoxynaphthalene
M2018-25G
M2018-5G
TCIAM2018-25GEA
0
CNY
TCIAM2018-25G
TCIAM2018-5G
2-Methoxy-1-naphthalenemethanol ≥98.0% (by GC)
2-Methoxy-1-naphthalenemethanol
分子式:
C₁₂H₁₂O₂ 分子量: 188.23 g/mol |
MDL编号:
MFCD00274224 CAS号: 40696-22-8 |
产品技术规格
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...