Megestrol acetate ≥96.0% (通过HPLC)

供应商: TCI
Warning

M1949-1G M1949-5G
TCIAM1949-1GEA 0 CNY
TCIAM1949-1G TCIAM1949-5G
Megestrol acetate ≥96.0% (通过HPLC)
Megestrol acetate
分子式: C₂₄H₃₂O₄
MDL编号: MFCD00056470
CAS号: 595-33-5
EINECS: 209-864-5
Merck Index: 14,05805

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 213.0 to 220.0 deg-C
Specific rotation [a]20/D +9.0 to +13.0 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR